Läs mer
Motic™ SMZ-168 Stereozoom™ Microscope
Produktkod. 11743215
Okular:
Binokulär
Trinokulär
Förpackningsstorlek:
Varje
Produktkod. 11743215
Brand: Motic™ PX660C6201
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Enables precise reproduction of magnifications with 5-position click-stop magnification mechanism implemented into the continuous zoom. Motic™ SMZ168 Series Stereo Zoom Microscope combines an impressive 3-dimensional upright image with an industry leading working distance of 113mm in its standard configuration to allow easy manipulation of the sample.
  • Greenough Optical System
  • Achromatic optics with anti-fungus treatment
  • Widefield binocular/trinocular 35° observation tube (light distribution 0:100 left eyetube/trinotube in photo position)
  • Widefield high eyepoint eyepieces, WF 10x23 with diopter adjustment on both eyepiece tubes ±5dpt
  • Interpupillary distance 52 to 75mm
  • Zoom: 6.7:1; 5 clickstops
  • 7.5X to 5X standard magnification
  • Working distance: 113mm
Specifikationer
Inkluderar Black, white and frosted stage plates and dust cover
Förstoringskraft 0.75X to 5X
Zoomområde Ratio 1:6,7
Modell SMZ168 BL
Huvud stil Rotating
Typ av belysning Halogen
Okular Binokulär