Learn More
Buerkle™ SteriBag™ StandUp Polyethylene Sampling Bags
Buerkle™ SteriBag™ StandUp Polyethylene Sampling Bags are sterile sample bag with a stand-up base.
1830.00 SEK - 2200.00 SEK
Specifications
Product Type | Sample Bag |
---|---|
Color | Clear |
For Use With (Application) | Ideal for Samples Available as Liquids, Powders, Solids and Pastes, Suitable for Foods and Can be Frozen |
Material | Polyethylene |
Sterility | Sterile |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17219549
|
Buerkle™
5344-0400 |
400 mL |
1830.00 SEK
Pack of 250 |
In Stock
Log in to see stock. |
|||||
15188136
|
Buerkle™
5344-0400 |
400 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15108146
|
Buerkle™
5344-0600 |
600 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Stands stably
- Protected against discharge
- Sterilised
- With labelling field
Specifications
Sample Bag | |
Ideal for Samples Available as Liquids, Powders, Solids and Pastes, Suitable for Foods and Can be Frozen | |
Sterile | |
Tight closing | |
229 mm |
Clear | |
Polyethylene | |
500 | |
500/Pk. | |
229 mm |