Läs mer
B Braun™ Sterican™ Intramuscular Cannula
Nålmätare:
14 G
Förpackningsstorlek:
Paket med 100
Beskrivning
- According to ISO 7864 and DIN 13097
- Thin-walled cannula
- Made of stainless chromium-nickel steel
- With extremely smooth surface and light silicone coating
- Special facet long grind
- For pain-free punctures
- Transparent Luer-Lock plastic hub made of polypropylene
- Colour coded according to ISO 6009
- Pack of 100
Specifikationer
Specifikationer
| Material | Stainless Chromium Nickel Steel |
| Färg | Light Green |
| Nålmätare | 14 G |
| Diameter (metrisk) Yttre | 2.1 mm |
| Längd (metrisk) | 80 mm |
| Produkttyp | Special Indication |
| Kvantitet | 1/Ea. |