missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sterilin™ Polystyrene Urine Tubes

Eliminate the need for decanting in the laboratory with Thermo Scientific™ Sterilin™ Polystyrene Urine Tubes. Ideal for use in automated urine analyzers.

Brand:  Thermo Scientific™ 131BBAC/C50

Additional Details : Weight : 7.34000kg

 View more versions of this product

Product Code. 15215619

  • 2653.32 SEK / Pack of 500

Estimated Shipment: 30-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Eliminate the need for decanting in the laboratory with Thermo Scientific™ Sterilin™ Polystyrene Urine Tubes. Ideal for use in automated urine analyzers.
NOT AVAILABLE IN AMERICA

Features:

  • Secure screw cap, leak-tested in accordance with BS EN 14254 Annexe D
  • Suitable for centrifugation at 3200xg
  • Aseptically manufactured
  • Fill level on the lable to ensure correct volume of sample submitted
  • Individually lot numbered for traceability

Note:

For in vitro use only.

Ordering Information:

Product is not available in all geographic regions.

Specifications

Specifications

0.43 oz.
13mL
Polystyrene w/PE closure
500
Polystyrene Urine Tube
Primary urine tube + boric acid, screw cap with collection cup
50
Aseptically Manufactured
Videos
SDS
Documents
Certificates