Läs mer
Thermo Scientific™ Sterilin™ Silicone Rubber Discs
Produktkod. p-6230192
Tjocklek (metrisk):
1,6 mm
3,2 mm
nr per ärende:
100
Förpackningsstorlek:
Paket med 100
Produktkod. 12812883
Brand: Thermo Scientific™ TDSC0220032
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Reduce opportunity for leakage in caps, injection ports and other applications. Translucent silicone material provides excellent performance over a wide temperature range.
Reduce opportunity for leakage in caps, injection ports and other applications. Translucent silicone material provides excellent performance over a wide temperature range. Thermo Scientific™ Sterilin™ Silicone Rubber Discs are suitable in a wide variety of applications.
  • Translucent
  • Operate over a wide temperature range: -50° to +200°C
  • Autoclavable
Ordering Information
  • Not available in all geographic regions
TRUSTED_SUSTAINABILITY
Specifikationer
Material Silicone Rubber
nr per ärende 100
Sterilitet Sterile
Temperaturområde (metriskt) -50° to +200°C
Tjocklek (metrisk) 3,2 mm
Produktlinje Sterilin
Form Round
Produkttyp Silicone Rubber Disc
Diameter (metrisk) 22 mm
Autoklaveringsbar Autoclavable