Learn More
Thermo Scientific™ Sterilin™ Test Tube Holders
Protect your test tubes and their contents with our flexible-cavity Thermo Scientific™ Sterilin™ Test Tube Holders. Unique design prevents them from floating in water baths.
901.80 SEK - 1927.80 SEK
Specifications
Material Rubber
View More Specs
Products 4
Product Code Brand Diameter (Metric) Opening Dimensions (L x W) Positions Holds Price Quantity & Availability  
Product Code Brand Diameter (Metric) Opening Dimensions (L x W) Positions Holds Price Quantity & Availability  
11319213
View Documents Promotion Details
Thermo Scientific™
BLKTTS4
25.4 mm 183 × 106mm 18 Tubes (25.4 mm) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11329213
View Documents Promotion Details
Thermo Scientific™
BLKTTS5
28.5 mm 180 × 105mm 15 Tubes (28.5 mm) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11339213
View Documents Promotion Details
Thermo Scientific™
BLKTTS6
9.4 mm 202 × 76mm 56 Tubes (9.4 mm) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11349213
View Documents Promotion Details
Thermo Scientific™
BLKTTS7
18.5 mm 183 × 105mm 28 Tubes (18.5 mm) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
Protect your test tubes and their contents with our flexible-cavity Thermo Scientific™ Sterilin™ Test Tube Holders. Unique design prevents them from floating in water baths.
  • Unique rubber design with flexible cavity diameters
  • Will sink in water baths and grips tubes to prevent them from floating to the surface
Ordering Information
  • Not available in all geographic regions
Specifications
Specifications
Rubber
Documents
Product Certifications
Note:
To ensure a secure grip, please select a cavity that is 1.5-2.0mm smaller than the tube diameter (e.g., 12.5mm cavity holds a 14mm diameter tube securely)