Learn More
Sampling Systems™ SteriWare™ Disposable Sample Spoons
Sampling Systems™ SteriWare™ Disposable Sampling Spoons are a range of low cost, single use spoons that allow for quick and efficient surface sampling of powders and granules.
655.00 SEK - 960.00 SEK
Product Code | Brand | Capacity (Metric) | Color | Length (Metric) | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Color | Length (Metric) | Sterility | Price | Quantity & Availability | |||
12782467
![]() |
Sampling Systems™
8088A-03S |
3 mL | White | 173 mm | Sterile |
915.00 SEK
Pack of 100 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||
15589634
![]() |
Sampling Systems™
8088R-10S |
10 mL | Red | 185 mm | Sterile |
960.00 SEK
Pack of 100 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||
15849461
![]() |
Sampling Systems™
8088A-03 |
3 mL | White | 173 mm | Non-sterile |
655.00 SEK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
15859461
![]() |
Sampling Systems™
8088A-10 |
10 mL | White | 185 mm | Non-sterile |
655.00 SEK
Pack of 100 |
Estimated Shipment: 06-08-2025
Log in to see stock. |
|||
15869461
![]() |
Sampling Systems™
8088B-10 |
10 mL | Blue | 185 mm | Non-sterile |
715.00 SEK
Pack of 100 |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||
15879461
![]() |
Sampling Systems™
8088B-10S |
10 mL | Blue | 185 mm | Sterile |
930.00 SEK
Pack of 100 |
Estimated Shipment: 18-08-2025
Log in to see stock. |
|||
12544366
![]() |
Sampling Systems™
8088A-10S |
10 mL | White | 185 mm | Sterile |
915.00 SEK
Pack of 100 |
Estimated Shipment: 01-08-2025
Log in to see stock. |
|||
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
Spoon | |
Single-use |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Sample Spoons