Läs mer
Sampling Systems™ SteriWare™ LiquiDip Mono
Handla allt Sampling Systems Produkter
Kapacitet (metrisk):
80 mL
Förpackningsstorlek:
Paket med 20
Beskrivning
- Assembled and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming HDPE
- BSE/TSE free
- Suitable for taking cross sectional samples
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- Available in 2 different lengths
- Push the sampler into the product, ensuring the sampler is in the open position
- Push the sampler against the base of the container to close the valve
- Remove the sampler and pour out the liquid into a suitable container
- No cross contamination
- No cleaning required
Specifikationer
Specifikationer
| Produkttyp | Liquid Sampler |
| Material | HDPE |
| Inkluderar | Valve at one end |
| Kapacitet (metrisk) | 80 mL |
| Disponibel | Single-use |
| Sterilitet | Non-sterile |
| Diameter (metrisk) Yttre | 21 mm |
| Längd (metrisk) | 450 mm |