Learn More
Kimberly-Clark™ KIMTECH SCIENCE™ STERLING™ NITRILE-XTRA™ Nitrile Gloves
Used for life sciences, forensic science and non-sterile drug manufacturing applications. Kimberly-Clark™ KIMTECH SCIENCE™ STERLING™ NITRILE-XTRA™ Nitrile Gloves provide protection and comfort in the laboratory.
192.61 SEK - 1833.40 SEK
Specifications
Color Gray
Classification PPE Category III
Finger Style Textured fingertips
Hand Ambidextrous
Inspected for Medical Use Yes
View More Specs
Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11788522 View Documents Kimberly-Clark™
98343
Medium 198.00 SEK
Pack of 100 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 117
Add to basket
11778522 View Documents Kimberly-Clark™
98342
Small 192.61 SEK
Pack of 100 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 135
Add to basket
11708532 View Documents Kimberly-Clark™
98345
X-Large 243.00 SEK
Pack of 90 View alternative products
Save Up to 
»
Remote Stock
(10 days availability): 28
Add to basket
11768522 View Documents Kimberly-Clark™
98341
X-Small 227.00 SEK
Pack of 100 View alternative products
Save Up to 
»
Sweden Stock
(immediate availability): 3
Add to basket
11798522 View Documents Kimberly-Clark™
98344
Large 198.00 SEK
Pack of 100 View alternative products
Save Up to 
»
Estimated Shipment
30-07-2019
Add to basket
11650403 View Documents Kimberly-Clark™
98342
Small This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Tested against over 40 chemicals and 10 cytotoxic drugs
  • Strength, quality and cleanliness tracked by certificate of analysis
  • PPE Category III compliant, AQL 0.65, EN374-1 and -2, ISO 16604, food contact approved
  • 30cm Powder free, ambidextrous gloves
  • Long beaded cuff for added strength
  • Textured fingertips for an improved grip
  • Available in sizes XS to XL
  • Silicone free formulation
  • Efficient materials and packaging can reduce waste by up to 33%
Specifications
Specifications
Gray
Textured fingertips
Yes
Nitrile
Ideal for use in life sciences, university labs, forensic science and non-sterile drug manufacturing
Yes
Beaded cuff
0.10mm Middle finger, 0.08mm palm, 0.07mm cuff
Gloves
PPE Category III
Ambidextrous
Yes
Puncture and abrasion, chemicals, Virus
Powder-free
300mm
38 to 42MPa
CE 0120 PPE Category III product classified by EC Council Directive 89/686/EEC, Tested in accordance with the EN Norms EN420:2003, Protection: EN374-1 EN374-2, s EN455-1,-2,-3,-4, ISO 16604 procedure B:2004, EN1149-5:2003, s AQL 0.65 G1, ISO 9001 and ISO 13485, FDA CFR 21 part 820