missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Steroidogenic Factor-1 (SF-1), Clone N1665, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Steroidogenic Factor-1 (SF-1), Clone N1665, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1946-P

Additional Details : Weight : 0.00250kg

Product Code. 15250665

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Streroidogenic Factor 1 (SF-1) is an orphan nuclear receptor belonging to the NR5A subgroup involved in gonadal and adrenal development. It may regulate genes by forming homodimers, heterdimers or as a monomer. A study by Y Tian et al suggests a significant correlation between the expression levels of SF-1 and StAR and stage of endometriosis in ectopic endometria; with higher levels observed in early stage. SF-1 is valuable in differential diagnosis of various types of ovarian tumors as well as in detecting endometrioid alterations2. SF-1 is a highly sensitive and specific differential diagnosis marker of ovarian sertoli cell tumors4. SF-1 expression is of stage- independent prognostic value in adrenocortical tumors and in determining adenocortical origin of an adrenal mass3. SF-1 plays a key role in the development of steroidogenic tissues, the regulatory of steroid biosynthesis and the pathogenesis of adrenocortical carcinoma5.

Host Species: Mouse
Clone: N1665
Isotype: IgG1
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: E.coli expressed recombinant SF-1 (220-461 aa)
Molecular Weight: ∼55kDa
Positive Control: Adrenal gland and spleen
Cellular Localization: Nuclear with occasional cytoplasmic
Recommended for: Immunohistochemistry
Specifications

Specifications

Immunohistochemistry
Monoclonal
Unconjugated
Mouse
∼55kDa
1 mL
Primary
Store vial at 4°C. When stored at 2-8°C, this antibody is stable for 24 months.
IgG1
Nuclear with occasional cytoplasmic
N1665
Antibody purified from ascites prepared in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
E.coli expressed recombinant SF-1 (220-461 aa)
Adrenal gland and spleen
RUO
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers