Läs mer
Steviol glycosides, TRC
Produktkod. 30416862
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30416862 1 g
1 options
Produktkod. 30416862 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6867301G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Stevioside
CAS 57817-89-7
Molekylformel C38 H60 O18
Kvantitet 1 g
Synonym Kaur-16-en-18-oic acid','13-[(2-O-β-D-glucopyranosyl-β-D-glucopyranosyl)oxy]-','β-D-glucopyranosyl ester','(4α)-, Stevioside (6CI,7CI), 1H-2,10a-Ethanophenanthrene','kaur-16-en-18-oic acid deriv., E 960, Stevian 50, Steviosin, α-G-Sweet
InChI formel InChI=1S/C38H60O18/c1-16-11-37-9-5-20-35(2,7-4-8-36(20,3)34(50)55-32-29(49)26(46)23(43)18(13-40)52-32)21(37)6-10-38(16,15-37)56-33-30(27(47)24(44)19(14-41)53-33)54-31-28(48)25(45)22(42)17(12-39)51-31/h17-33,39-49H,1,4-15H2,2-3H3/t17-,18-,19-,20+,21+,22-,23-,24-,25+,26+,27+,28-,29-,30-,31+,32+,33+,35-,36-,37-,38+/m1/s1
LEDER C[C@@]12CCC[C@](C)([C@H]1CC[C@]34CC(=C)[C@](CC[C@@H]23)(C4)O[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)C(=O)O[C@@H]7O[C@H](CO)[C@@H](O)[C@H](O)[C@H]7O
Molekylvikt (g/mol) 804.87
Formel vikt 804.378