Läs mer
Hanna Instruments™ PICCOLO™ pH Tester with 3.5 in. Probe
Mått (L x B x H):
194 x 29 x 15 mm
Förpackningsstorlek:
Varje
Beskrivning
The PICCOLO™ is a lightweight digital tester that is easy to use with a replaceable probe. Conventional pH meters are susceptible to the weak, high impedance signal which makes the electrode, connector, cable and meter vulnerable to noise, humidity and dirty environments. PICCOLO has overcome these problems with a pre-amplified electrode delivering a strong signal to the meter.
- Replaceable, pre-amplified 3.5 in. electrode (HI1280)
- Automatic Temperature Compensation (ATC)
- Easy two-point manual calibration
- 100 hour battery life
- Simple to use
- Economical
Specifikationer
Specifikationer
| Inkluderar | 90mm (3.5 in.) HI1280 amplified pH electrode, pH 4.01 and pH 7.01 buffer solution sachets, calibration screwdriver, batteries, rugged carrying case and instructions |
| Noggrannhet (pH) | ±0.01 |
| Kalibrering | Manual |
| Område (pH) | 1.00 to 13.00 |
| Upplösning (pH) | 0.01 |
| Upplösning (mV) | 0.1mV |
| Kalibreringspunkter | 2 |
| Mått (L x B x H) | 194 x 29 x 15 mm |
| Vikt (metrisk) | 70 g |
| Vikt (engelska) | 2.5 oz. |
| Visa mer |