Läs mer
Stigmasteryl Ferulate, TRC
Produktkod. 30415026
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30415026 2.5 mg 2.5mg
30416611 100 mg -
30413399 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30415026 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6867682.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20972-08-1
Molekylformel C39H56O4
Molekylvikt (g/mol) 588.86
Synonym Stigmasterol Ferulate, 4-hydroxy-3-methoxy-stigmasta-5,22-dien-3β-yl Ester Cinnamic Acid, 4-Hydroxy-3-methoxycinnamate Stigmasterol, (3β,22E)-3-(4-Hydroxy-3-methoxyphenyl)-2-propenoate Stigmasta-5,22-dien-3-ol
IUPAC-namn [(3S,8S,9S,10R,13R,14S,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] 3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
LEDER O=C(/C=C/C1=CC(OC)=C(O)C=C1)O[C@@H]2CC3=CC[C@]4([H])[C@@](CC[C@@]5(C)[C@@]4([H])CC[C@]5([H])[C@H](C)/C=C/[C@@H](CC)C(C)C)([H])[C@]3(CC2)C
Specifikationer
Kemiskt namn eller material Stigmasteryl Ferulate
Kvantitet 2.5 mg
InChI formel InChI=1S/C39H56O4/c1-8-28(25(2)3)12-9-26(4)32-15-16-33-31-14-13-29-24-30(19-21-38(29,5)34(31)20-22-39(32,33)6)43-37(41)18-11-27-10-17-35(40)36(23-27)42-7/h9-13,17-18,23,25-26,28,30-34,40H,8,14-16,19-22,24H2,1-7H3/b12-9+,18-11?/t26-,28-,30+,31+,32-,33+,34+,38+,39-/m1/s1
Formel vikt 588.4179