Läs mer
Stiripentol, TRC
Produktkod. 30009917
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009917 250 mg
17923375 50 mg
17951775 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009917 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS686825250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 49763-96-4
Molekylformel C14 H18 O3
Molekylvikt (g/mol) 234.29
Synonym 1-Penten-3-ol, 1-(1,3-benzodioxol-5-yl)-4,4-dimethyl-, 1-(1,3-Benzodioxol-5-yl)-4,4-dimethyl-1-penten-3-ol, 4,4-Dimethyl-1-(3,4-methylenedioxyphenyl)-1-penten-3-ol, BCX 2600, Diacomit, Stiripentol
IUPAC-namn (E)-1-(1,3-benzodioxol-5-yl)-4,4-dimethylpent-1-en-3-ol
LEDER CC(C)(C)C(O)\C=C\c1ccc2OCOc2c1
Specifikationer
Kemiskt namn eller material Stiripentol
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C14H18O3/c1-14(2,3)13(15)7-5-10-4-6-11-12(8-10)17-9-16-11/h4-8,13,15H,9H2,1-3H3/b7-5+
Formel vikt 234.1256
Procent renhet >95%