missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK16 (PKL12) Recombinant Human Protein
STK16 (PKL12) is a serine/threonine kinase.
Brand: Invitrogen™ PR7859A
Additional Details : Weight : 0.01000kg
Description
STK16 (PKL12) functions in signal transduction pathways and transcriptional regulation.
Specifications
NP_003682 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
100μg | |
RUO - research use only | |
STK16 (PKL12), MPSK, TSF1 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
38.7kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
NAK Kinase Family | |
Recombinant | |
Baculovirus |