missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STK22D (TSSK1) Recombinant Human Protein
STK22D (TSSK1) is a serine/threonine kinase.
18130.00 SEK - 79000.00 SEK
Specifications
Accession Number | NP_114417 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl, pH 7.5, 100mM NaCl, 0.5mM EDTA, 0.05% NP40, 4mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 46.7kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468433
|
Invitrogen™
PR5214A |
100μg |
18130.00 SEK
100µg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10492303
|
Invitrogen™
PV4063 |
1mg |
79000.00 SEK
1mg |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
STK22D (TSSK1) is essential to the function and production of sperm (spermiogenesis).
Specifications
NP_114417 | |
50mM Tris-HCl, pH 7.5, 100mM NaCl, 0.5mM EDTA, 0.05% NP40, 4mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
TSSK Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
46.7kDa | |
RUO - research use only | |
STK22D (TSSK1) | |
Human | |
His Tag | |
Frozen |