Learn More
Invitrogen™ STK32B (YANK2) Recombinant Human Protein
STK32B or YANK2 is a serine/threonine protein kinase that contains motif for binding metal ions and nucleotides. Full-length recombinant human STK32B (YANK2) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Brand: Invitrogen™ PV6408
Additional Details : Weight : 0.01000kg
Description
The gene for STK32B is highly conserved among various species. STK32B has been associated with isolated cleft lip with or without cleft palate and cleft palate which are among the most common human birth defects of oral cleft cases. Long interspersed nuclear element-1 (LINE-1 or L1) mediated deletion of STK32B gene is observed in patients with Ellis-van Creveld syndrome with borderline intelligence.
- Validated in LanthaScreen™ Kinase Binding Assay
- Protocols available for simple use and optimized performance
- Multiple pack sizes available for convenient use
Specifications
Enzyme and Protein Activity Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Kinase Proteins and Expression Reagents, Pharma and Biopharma, Protein Assays and Analysis, Protein Biology, Target and Lead Identification and Validation | |
74.1kDa | |
Store at -68 to -85°C. Aliquot at first use to avoid multiple freeze-thaws. | |
Contains R198G SNP | |
Recombinant |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.25mM DTT, 10mM glutathione, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol | |
1mg | |
YANK2, STK32B, STK32, STKG6, HSA250839 | |
Human | |
GST Tag |