missing translation for 'onlineSavingsMsg'
Learn More

STK32C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 11434-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16828513

  • 1656.73 SEK / 20µL
Estimated Shipment: 03-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the serine/threonine protein kinase family. The specific function of this kinase is not known.
Specifications

Specifications

STK32C
Polyclonal
Unconjugated
STK32C
PKE, RP11 140A10.1, serine/threonine kinase 32C, STK32C, YANK3
Rabbit
Antigen Affinity Chromatography
RUO
282974, 365381, 57740
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q86UX6, Q8QZV4
STK32C
STK32C Fusion Protein Ag1960
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers