missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK32C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | STK32C |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828513
|
Proteintech
11434-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818513
|
Proteintech
11434-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. The specific function of this kinase is not known.Specifications
STK32C | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q86UX6, Q8QZV4 | |
282974, 365381, 57740 | |
STK32C Fusion Protein Ag1960 | |
Primary | |
-20°C | |
STK32C |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PKE, RP11 140A10.1, serine/threonine kinase 32C, STK32C, YANK3 | |
STK32C | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |