missing translation for 'onlineSavingsMsg'
Learn More

STK38L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12697-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880554

  • 3665.00 SEK / 150µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody is specific to STK38L.

STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal. Northern blot analysis revealed expression of a 3.9-kb transcript in all tissues tested, with the possible exception of adult brain. Highest expression was in peripheral blood leukocytes. Immunoblot and immunofluorescence microscopy demonstrated predomitly nuclear expression of a 55-kD protein.
Specifications

Specifications

STK38L
Polyclonal
Unconjugated
STK38L
KIAA0965, NDR2, NDR2 protein kinase, Nuclear Dbf2 related kinase 2, STK38L
Rabbit
Antigen Affinity Chromatography
RUO
23012, 232533, 691337
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7TSE6, Q9Y2H1
Stk38l
STK38L Fusion Protein Ag3413
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers