Läs mer
Invitrogen™ STMN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA591899
Beskrivning
Immunogen sequence: MTLAAYKEKM KELPLVSLFC SCFLADPLNK SSYKYEGWCG RQCRRKDESQ RKDSADWRER RAQADTVDLN WCVISDMEVI ELNKCTSGQS FEVILKPPSF DGVPEFNASL PRRRDPSLEE IQKKLEAAEE RRKYQEAELL KHLAEKREHE REVIQKAIEE NNNFIKMAKE KLAQKMESNK ENREAHLAAM LERLQEKDKH AEEVRKNKEL KEEASR; Positive Samples: Mouse spinal cord, Rat spinal cord; Cellular Location: Cell projection, Golgi apparatus, axon, growth cone.
RB3 (Stathmin-4), encoded by STMN4 gene, is a neuron-specific homologue of the SCG10/stathmin family proteins, possessing a unique N-terminal membrane-associated domain and the stathmin-like domain at the C terminus, which promotes microtubule (MT) catastrophe and/or tubulin sequestering. Stathmin is a ubiquitous cytosolic phosphoprotein, preferentially expressed in the nervous system, and described as a relay integrating diverse intracellular signaling pathways. RB3 is consistently up-regulated in differentiated neuroblastoma cells, and stable expression of transfected RB3 could induce differentiation, suggesting a role of RB3 during neuroblastoma differentiation. Moreover, increased RB3 is a unique response protein in the stathmin-related proteins following optical nerve axotomy and RB3 may be a critical target for neurodegenerative diseases.
Specifikationer
| STMN4 | |
| Polyclonal | |
| Unconjugated | |
| STMN4 | |
| Lagl; R75359; Rb3; stathmin 4; stathmin-4; stathmin-like 4; stathmin-like protein B3; stathmin-like protein RB3; stathmin-like-protein RB3; Stmn4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56471, 79423, 81551 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P63042, P63043, Q9H169 | |
| STMN4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-216 of human STMN4 (NP_1104222). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.