Läs mer
Tocris Bioscience™ STOCK2S 26016, >98%
GREENER_CHOICE
Produktkod. 30002144
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30002144 10 mg
30000748 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30002144 Leverantör Tocris Bioscience™ Leverantörsnummer 5570/10
Lysine deficient protein kinase (WNK) signaling inhibitor
Lysine deficient protein kinase (WNK) signaling inhibitor (IC50 = 16 μM). Inhibits binding of WNK1 and WNK4 to SPAK and phosphorylation of downstream transporters, NKCC1 and NCC, in mouse vascular smooth muscle and distal convoluted tubule cell cultures exposed to hypotonic shock, respectively.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 332922-63-1
Molekylformel C20H19N3O2
Specifikationer
Kemiskt namn eller material 7-Ethoxy-N3-(2-furanylmethyl)-3,9-acridinediamine
Kvantitet 10 mg
Löslighet Soluble to 100mM in DMSO and to 50mM in ethan
Formel vikt 333.38
Renhet >98%
Molekylvikt (g/mol) 333.391