Läs mer
Stomatin Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67894-1-IG-20UL
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
The Stomatin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Stomatin. This antibody recognizes human antigen. The Stomatin antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifikationer
| Stomatin | |
| Monoclonal | |
| 1000 μg/mL | |
| BND7; EPB7; EPB72 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 32 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, ELISA | |
| 1A4D6 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 2040 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |