Learn More
Carl Zeiss™ Stop Slider A, 14 x 40mm
Features an aperture diaphragm for Axio Scope and a luminous field diaphragm for Axio Vert Microscope
Brand: Carl Zeiss™ 423730-9151-000
Additional Details : Weight : 1.00000kg
Description
Compatible Microscopes:
- Axio Scope.A1
- Axio Scope.A1 Pol
- Axio Scope.A1 Vario
- Axio Vert.A1 FL
- Axio Vert.A1 MAT
Specifications
Stop Slider A with Aperture/Luminous Field Diaphragm | |
Axio Scope; Axio Vert Microscopes | |
Aperture/luminous field diaphragm |