Learn More
Lenz Laborglasintrumente Stoppers Octagonal Plate, Plastic
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
61.85 SEK - 131.44 SEK
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10010361
|
Lenz Laborglasintrumente
01401034 |
NS 34/35 |
131.44 SEK
Pack of 5 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10224531
|
Lenz Laborglasintrumente
01401010 |
NS 10/19 |
79.61 SEK
Pack of 10 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
10643622
|
Lenz Laborglasintrumente
01401007 |
NS 7/16 |
61.85 SEK
Pack of 10 |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||||
Description
- Acc. to DIN 12252
- Form A and E
- Solid or semi-hollow
- Material: Plastic
Specifications
White, Red | |
Polyethylene |