Learn More
BRAND™ Storage Box For Cryogenic Tubes
Holds 81 tubes
Brand:  BRAND™ 114864
1423.59 SEK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.90000kg
 View more versions of this product
Product Code. 15501281
  • 1650.00 SEK / Pack of 5
In Stock
to see stock.
Add to basket
Description
Description
  • Polycarbonate (PC)
  • For cryogenic tubes
  • Operating range (in the gaseous phase of liquid nitrogen) -196 °C to +121 °C
  • Autoclavable at 121 °C (2 bar)
  • DIN EN 285
  • Specifications
    Specifications
    Polycarbonate
    3 mL, 4 mL and 5 mL cryogenic tubes
    5
    Cryo Storage Box
    81 (3 mL, 4 mL and 5 mL Cryogenic Tubes)