Learn More
Lenz Laborglasinstrumente™ Straight Adapter
Lenz Laborglasinstrumente™ Adapters are manufactured from durable DURAN™ glass tubing.
Brand: Lenz Laborglasinstrumente™ 05101004
Additional Details : Weight : 0.80000kg
Description
- Straight glass olive
- Available in a range of joint sizes
Specifications
Straight Adapter | |
Duran Glass | |
NS 29/32 | |
Socket |