Learn More
BRAND™ Straight Burette Stopcock and Spare Key, Glass
Straight Burette Stopcock and Spare Key is suitable for use with 25 mL and 50 mL burettes.
Brand:  BRAND™ 81247
Additional Details : Weight : 0.02000kg
Product Code. 10089850
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Stopcock housing with burette tip
  • Interchangeable PTFE or borosilicate glass key with retention device
Specifications
Specifications
Straight Burette Stopcock
Borosilicate Glass (3.3)
25 and 50 mL Burette
Spare Key, Securing Device