Learn More
Fisherbrand™ Straight Sided Jars with White Polypropylene Caps
Catalog Number p-7145769
Click to view available options
Capacity (English):
1 oz.
2 oz.
8 oz.
16 oz.
32 oz.
Capacity (Metric):
30 mL
60 mL
240 mL
480 mL
950 mL
Fisherbrand™ Straight Sided Jars with White Polypropylene Caps are ideal for large solid samples and allow for complete removal of contents.
  • Translucent and durable
  • Jars are ideal for storing and collecting specimens and lab supplies
  • Wide neck and straight sides facilitate easy recovery of contents
  • Polypropylene offers greater tolerance to high temperatures than HDPE and better contact clarity
  • Features wide neck and straight sides to facilitate easy recovery of contents
Closure Material Polypropylene
Closure Size 43-400 mm
Closure Type Cap
Diameter (Metric) 38 mm
Height (Metric) 40 mm
Product Type Straight Sided Jar
Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Natural
Diameter (Metric) Outer Neck 39 mm
Diameter (Metric) Inner Neck 37 mm
For Use With (Application) Large Solid Samples and Allow for Complete Removal of Contents
Includes Cap
Height (Metric) With Closure 43 mm
Material Polypropylene
Quantity 72
Closure Color White
Neck Finish 43-400
Clarity Translucent
Closure Included Yes
Mouth Wide
Shape Round