Beskrivning
- Available retention mechanisms include reversed phase, ion exchange, normal phase and mixed mode
- 24 phases and counting
- Cleaner extracts and reproducible results
Specifikationer
Specifikationer
| För användning med (utrustning) | Vacuum Manifolds or Automated SPE Equipment |
| Partikelstorlek | 25 μm |
| Sorbent | C8 |
| Typ | Solid Phase Extraction Column |