missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STRBP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | STRBP |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827244
|
Proteintech
17362-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817244
|
Proteintech
17362-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 22-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Involved in spermatogenesis and sperm function. Plays a role in regulation of cell growth. Binds to double-stranded DNA and RNA. Binds most efficiently to poly(I:C) RNA than to poly(dI:dC) DNA. Binds also to single-stranded poly(G) RNA. Binds non-specifically to the mRNA PRM1 3′-UTR and adenovirus VA RNA.Specifications
STRBP | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q91WM1, Q96SI9, Q9JKU6 | |
20744, 55342, 84476 | |
STRBP Fusion Protein Ag10720 | |
Primary | |
-20°C | |
STRBP |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ILF3L, p74, SPNR, STRBP | |
STRBP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |