Learn More
IBA GmbH™ Strep-Tactin™ Chromeo 546 Conjugate
IBA GmbH™ Strep-Tactin™ Chromeo 546 Conjugate is for detection of Strep-tag™ and Twin-Strep-tag™ fusion proteins.
Brand:  IBA GmbH™ 2-1548-050
Additional Details : Weight : 0.01000kg
Product Code. 15332016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • For the detection of Strep-tag™ II and Twin-Strep-tag™ in Immunofluorescence, FACS and more
  • Contains Strep-Tactin™ conjugated to Chromeo 546
  • 1 x 50μg fluorescent conjugate
Specifications
Specifications
50μg