Läs mer
IBA Lifesciences Strep-Tactin™XT Superflow™ resin
Produktkod. 15533077 Handla allt IBA Lifesciences Produkter
Change view
Kapacitet (metrisk):
4 mL
20 mL
50 mL
1000 mL
Förpackningsstorlek:
1000ml
20ml
4ml
50ml
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kapacitet (metrisk) unitSize
15533077 1000 mL 1000ml
15513077 20 mL 20ml
15523077 50 mL 50ml
15583756 4 mL 4ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 15533077 Leverantör IBA Lifesciences Leverantörsnummer 24010500
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Strep-Tactin™XT Superflow™ resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™XT Superflow™ is a 6% agarose coupled with the streptavidin variant Strep-Tactin™XT and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • As with any other Strep-Tactin™XT resins, the unspecific protein binding is extremely low, which results in an outstanding purity of the target protein.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™XT Superflow™ has a higher affinity for Strep-tag™ fusion proteins, which leads to an improved immobilization and purification.
  • In addition, Strep-Tactin™XT Superflow™ allows the application of denaturing agents such as urea during the purification procedure.
  • The suspension is suitable for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension are necessary.
Specifikationer
Kapacitet (metrisk) 1000 mL
Typ Resin