Läs mer
Streptamine Sulfate, TRC
Produktkod. 30422617
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30422617 25 mg
30420506 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30422617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS68735025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5469-74-9
Molekylformel C6H14N2O4 .H2O4S
Molekylvikt (g/mol) 276.27
Synonym Streptamine, Sulfate (1:1) (8CI,9CI)
IUPAC-namn (1R,3S,4R,6S)-4,6-diaminocyclohexane-1,2,3,5-tetrol;sulfuric acid
LEDER N[C@@H]1[C@@H](O)[C@H](N)[C@@H](O)[C@H](O)[C@H]1O.OS(=O)(=O)O
Specifikationer
Kemiskt namn eller material Streptamine Sulfate
Kvantitet 25 mg
InChI formel InChI=1S/C6H14N2O4.H2O4S/c7-1-3(9)2(8)5(11)6(12)4(1)10;1-5(2,3)4/h1-6,9-12H,7-8H2;(H2,1,2,3,4)/t1-,2+,3-,4+,5-,6-;
Formel vikt 276.0627