missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (S69S)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518262
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is reactive with the S. pneumoniae common C-polysaccharide (backbone epitope). Purity is >90%.
There are more than 85 antigenic types of S. pneumoniae, which are determined by capsule antigens. Altough S. pneumoniae is part of the normal respiratory flora, invasion commonly results in pneumonia as well as meningitis. S. pneumoniae is protected from phagocytosis by its polysaccharide capsule which is also a prominent virulence factor. Infections can usually be treated with antibiotics, although resistant strains are emerging.
Specifikationer
| Streptococcus pneumoniae | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG3 |
| ELISA, Immunocytochemistry | |
| S69S | |
| Unconjugated | |
| Pneumococcus; s pneumoniae; S. pneumoniae | |
| Strep pneumoniae native protein. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering