Learn More
Strontium Chloride Hexahydrate, Supelco™
Product Code. 15895060
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15895060 1 kg 1kg
15857620 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15895060 Supplier Supelco Supplier No. 1078651000
 Restricted Item
Beräknad leverans: 25-11-2026
For analysis EMSURE™ ACS
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10025-70-4
Specifications
Melting Point 61°C
Quantity 1 kg
MDL Number MFCD00149865
Synonym Strontium chloride hexahydrate, Strontium chloride hexahydrate
Molecular Weight (g/mol) 266.62
Grade Research
Density 1.95 g/cm3 at 20°C
Chemical Name or Material Strontium Chloride Hexahydrate