Learn More
Strontium Chloride, TRC
Specifikationer
Specifikationer
| Chemical Name or Material | Strontium Chloride |
| CAS | 10476-85-4 |
| Molecular Formula | SrCl2 |
| Quantity | 100 g |
| Synonym | Strontium chloride (6CI,8CI), STC-K, Strontium Dicholoride |
| InChI Formula | InChI=1S/2ClH.Sr/h2*1H;/q;;+2/p-2 |
| SMILES | [Cl-].[Cl-].[Sr+2] |
| IUPAC Name | strontium;dichloride |
| Molecular Weight (g/mol) | 158.53 |
| Formula Weight | 157.8433 |