Läs mer
Strophanthidol, TRC
Produktkod. 30424224
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424224 10 mg
30411716 50 mg
30422736 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424224 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS56054010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 560-54-3
Molekylformel C23 H34 O6
Molekylvikt (g/mol) 406.51
Synonym (3β,5β)-3,5,14,19-Tetrahydroxy-card-20(22)-enolide, k-Strophanthidol
IUPAC-namn 3-[(3S,5S,10R,13R,14S,17R)-3,5,14-trihydroxy-10-(hydroxymethyl)-13-methyl-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one
LEDER C[C@]12CCC3C(CC[C@]4(O)C[C@@H](O)CC[C@]34CO)[C@@]1(O)CC[C@@H]2C5=CC(=O)OC5
Specifikationer
Kemiskt namn eller material Strophanthidol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C23H34O6/c1-20-6-3-17-18(4-8-22(27)11-15(25)2-7-21(17,22)13-24)23(20,28)9-5-16(20)14-10-19(26)29-12-14/h10,15-18,24-25,27-28H,2-9,11-13H2,1H3/t15-,16+,17?,18?,20+,21-,22-,23-/m0/s1
Formel vikt 406.2355
Procent renhet >95%