Läs mer
Strychnine Nitrate, TRC
Produktkod. 30425476
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425476 500 mg
30421271 2.5 g
30411184 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425476 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS687235500MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66-32-0
Molekylformel C21 H22 N2 O2 . H N O3
Molekylvikt (g/mol) 397.42
Synonym Strychnine Nitrate, Strychnidin-10-one, mononitrate (9CI), Strychnine, mononitrate (8CI), 4,6-Methano-6H,14H-indolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline, strychnidin-10-one deriv., Strychnine nitrate, Strychnidin-10-one, nitrate (1:1)
IUPAC-namn (4aR,5aS,8aR,13aS,15aS,15bR)-4a,5,5a,7,8,13a,15,15a,15b,16-decahydro-2H-4,6-methanoindolo[3,2,1-ij]oxepino[2,3,4-de]pyrrolo[2,3-h]quinoline-14-one;nitric acid
LEDER O[N+](=O)[O-].O=C1C[C@@H]2OCC=C3CN4CC[C@@]56[C@@H]4C[C@@H]3[C@@H]2[C@@H]5N1c7ccccc67
Specifikationer
Kemiskt namn eller material Strychnine Nitrate
Kvantitet 500 mg
InChI formel InChI=1S/C21H22N2O2.HNO3/c24-18-10-16-19-13-9-17-21(6-7-22(17)11-12(13)5-8-25-16)14-3-1-2-4-15(14)23(18)20(19)21;2-1(3)4/h1-5,13,16-17,19-20H,6-11H2;(H,2,3,4)/t13-,16-,17-,19-,20-,21+;/m0./s1
Formel vikt 397.1638