Läs mer
Cole-Parmer™ Stuart™ SCT1 Temperature Controller
Produktkod. 12008323
Change view
Noggrannhet:
±0,5 °C
Förpackningsstorlek:
Varje
Produktkod. Noggrannhet unitSize
12008323 ±0,5 °C Varje
1 options
Produktkod. 12008323 Leverantör Cole-Parmer™ Leverantörsnummer 0480592
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Designed for use with the Stuart™ Undergrad range of stirrer hotplates, the SCT1 can be used either as a temperature controller or as a digital thermometer. Stuart™ SCT1 Temperature Controller is ideal for the accurate temperature control of aqueous or oil based samples in the laboratory.
  • Accurate liquid temperature control
  • Built in retort fitting and probe holder
  • Detachable temperature probe
  • Bright, easy to read LED display
  • Compact and light weight
  • Comprehensive range of accessories
Specifikationer
Noggrannhet ±0,5 °C
Höjd (metrisk) 123 mm
Bredd (metrisk) 90 mm
Modell SCT1
Sensortyper PT100