missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STX12 Monoclonal Antibody (4E212)

Product Code. 11559422
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
50 μg
missing translation for 'unitSize'
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet unitSize
11559422 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11559422 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' MA126001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MA1-26001 detects a band of ~38 kDa corresponding to recombinant rat syntaxin 13. Suggested positive controls include rat or mouse brain extract.

SNARE that acts to regulate protein transport between late endosomes and the trans-Golgi network. The SNARE complex containing STX6, STX12, VAMP4 and VTI1A mediates vesicle fusion (in vitro).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX12
Användningsområden Immunohistochemistry (Paraffin), Western Blot
Klassificering Monoclonal
Klona '4E212
Koncentration 1 mg/mL
Konjugera Unconjugated
Formulering PBS with 50% glycerol and 0.09% sodium azide
Gen STX12
Genaccessionsnr. G3V7P1, Q86Y82, Q9ER00
Gene Alias AI850350; AU041521; MGC51957; Stx12; Stx13; STX14; Syn13; syntaxin 12; syntaxin 13; Syntaxin-12; Syntaxin-13
Gensymboler STX12
Värd art Mouse
Immunogen Recombinant cytoplasmic domain of rat syntaxin 13.
Reningsmetod Protein G
Kvantitet 50 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 100226, 23673, 65033
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.