missing translation for 'onlineSavingsMsg'
Learn More

Styrene, 99.5% stab. with 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog Number p-7049613
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy
100-42-5
C8H8
104.15
MFCD00008612,MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
C=CC1=CC=CC=C1
This item is not returnable. View return policy
100-42-5
C8H8
104.15
MFCD00008612,MFCD00084450
PPBRXRYQALVLMV-UHFFFAOYSA-N
ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
7501
CHEBI:27452
C=CC1=CC=CC=C1

Styrene is used in the preparation of polystyrene, which finds application in rigid foodservice containers, automobile body panels, wind turbine parts, boats, compact disc (CD) cases, envelope windows and many other products. Other uses include in refrigerator liners, in medical devices and in small household appliances. It is used as a precursor for the preparation of styrene-butadiene rubber, styrene-butadiene latex, styrene-isoprene-styrene (SIS), styrene-divinylbenzene (S-DVB) and styrene-acrylonitrile resin (SAN) which are used in rubber, plastic, insulation, fiberglass, pipes, automobile and boat parts, food containers and carpet backing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -31°C
Color Colorless
Density 0.906 g/mL
pH neutral
Boiling Point 145°C to 146°C
Flash Point 31°C (88°F)
Odor Pungent
Refractive Index 1.546
Quantity 50 mL
UN Number UN2055
Beilstein 1071236
Sensitivity Air and light sensitive
Merck Index 14,8860
Solubility Information Miscible with alcohols,ethers and carbon disulfide. Immiscible with water.
IUPAC Name styrene
Formula Weight 104.15
Percent Purity 99.5%
Grade Technical
Physical Form Liquid
Chemical Name or Material Styrene, Stabilized with 4-tert-butylcatechol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'