Läs mer
SU-5402 Ethyl Ester, TRC
Produktkod. 30425755
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30425755 5 mg
1 options
Produktkod. 30425755 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6880155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)methyl-4-methyl-1H-pyrrole-3-propanoic Acid Ethyl Ester
CAS 210644-65-8
Molekylformel C19H20N2O3
Kvantitet 5 mg
Synonym SU-5402 Ethyl Ester, 2-(1,2-Dihydro-2-oxo-3H-indol-3-ylidene)methyl-4-methyl-1H-pyrrole-3-propanoic Acid Ethyl Ester
InChI formel InChI=1S/C19H20N2O3/c1-3-24-18(22)9-8-13-12(2)11-20-17(13)10-15-14-6-4-5-7-16(14)21-19(15)23/h4-7,10-11,20H,3,8-9H2,1-2H3,(H,21,23)/b15-10-
LEDER CCOC(=O)CCc1c(C)c[nH]c1\C=C\2/C(=O)Nc3ccccc23
IUPAC-namn ethyl 3-[4-methyl-2-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoate
Molekylvikt (g/mol) 324.37
Formel vikt 324.1474