Learn More
SU 5402, TRC
Product Code. 30422057
Change view
Click to view available options
Kvantitet:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30422057 25 mg
30410544 1 mg
30420896 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30422057 Supplier Toronto Research Chemicals Supplier No. TRCS68800025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 215543-92-3
Molekylformel C17 H16 N2 O3
Molekylvikt (g/mol) 296.32
IUPAC-namn 3-[4-methyl-2-[(Z)-(2-oxo-1H-indol-3-ylidene)methyl]-1H-pyrrol-3-yl]propanoic acid
LEDER Cc1c[nH]c(\C=C\2/C(=O)Nc3ccccc23)c1CCC(=O)O
Specifications
Kemiskt namn eller material SU 5402
Kvantitet 25 mg
InChI formel InChI=1S/C17H16N2O3/c1-10-9-18-15(11(10)6-7-16(20)21)8-13-12-4-2-3-5-14(12)19-17(13)22/h2-5,8-9,18H,6-7H2,1H3,(H,19,22)(H,20,21)/b13-8-
Formel vikt 296.1161