Läs mer
SU 5416, TRC
Produktkod. 30428478
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428478 100 mg
30410737 50 mg
30409489 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428478 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS688030100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 204005-46-9
Molekylformel C15 H14 N2 O
Molekylvikt (g/mol) 238.28
Synonym 3-[(3,5-Dimethyl-1H-pyrrol-2-yl)methylene]-1,3-dihydro-2H-indol-2-one, 3-(3,5-Dimethyl-1H-pyrrol-2-ylmethylene)-1,3-dihydroindol-2-one, 3-[(3,5-Dimethylpyrrol-2-yl)methylene]indolin-2-one, NSC 696819, Semaxinib, Semoxind, Sugen 5416
IUPAC-namn (3Z)-3-[(3,5-dimethyl-1H-pyrrol-2-yl)methylidene]-1H-indol-2-one
LEDER Cc1cc(C)c(\C=C\2/C(=O)Nc3ccccc23)[nH]1
Specifikationer
Kemiskt namn eller material SU 5416
Kvantitet 100 mg
InChI formel InChI=1S/C15H14N2O/c1-9-7-10(2)16-14(9)8-12-11-5-3-4-6-13(11)17-15(12)18/h3-8,16H,1-2H3,(H,17,18)/b12-8-
Formel vikt 238.1106