Läs mer
Tocris Bioscience™ SU 6656
Beskrivning
SU 6656 is a potent ATP-competitive Src family kinase inhibitor (IC50 values are 0.02, 0.13, 0.17, 0.28 and 6.88 nM for Yes, Lyn, Fyn, Src and Lck, respectively). Exhibits > 6.5 fold selectivity for Src family over other kinases. Also AMPK inhibitor (IC50 = 0.22 μM for the α2 kinase domain). Cell permeable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 2,3-Dihydro-N,N-dimethyl-2-oxo-3-[(4,5,6,7-tetrahydro-1H-indol-2-yl)methylene]-1H-indole-5-sulfonamide |
| CAS | 330161-87-0 |
| Kvantitet | 25 mg |
| Molekylformel | C19H21N3O3S |
| Formel vikt | 371.45 |
| Renhet | 98% |