Läs mer
SU5408, MedChemExpress
Produktkod. 16496487
Encompass_Preferred
Change view
Kvantitet:
1 mg
5 mg
10 mg
25 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496487 5 mg
16462846 25 mg
16453768 10 mg
16430989 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496487 Leverantör Medchem Express Leverantörsnummer HY103002
MedChemExpress SU5408 (VEGFR2 Kinase Inhibitor I) is a potent and cell-permeable inhibitor of VEGFR2 kinase with an IC50 of 70 nM.
SU5408 (VEGFR2 Kinase Inhibitor I) is a potent and cell-permeable inhibitor of VEGFR2 kinase with an IC50 of 70 nM.
Specifikationer
Kemiskt namn eller material SU5408
Fysisk form Solid
CAS 15966-93-5
Färg Orange
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C18H18N2O3
För användning med (applikation) COVID-19-immunoregulation
Synonym VEGFR2 Kinase Inhibitor I
Löslighetsinformation DMSO : 6 mg/mL (19.33 mM; Need ultrasonic and warming)
LEDER O=C(C1=C(C)NC(/C=C2C(NC3=C\2C=CC=C3)=O)=C1C)OCC
Molekylvikt (g/mol) 310.35
Formel vikt 310.35
Procent renhet 98.0%