Läs mer
Suberanilic Acid, TRC
Produktkod. 30426101
Change view
Kvantitet:
100 mg
200 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30426101 250 mg
30417258 200 mg
30414345 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30426101 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS688650250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 149648-52-2
Molekylformel C14 H19 N O3
Molekylvikt (g/mol) 249.31
Synonym Octanoic acid, 8-oxo-8-(phenylamino)-, 8-Oxo-8-(phenylamino)octanoic acid, Suberanilic acid
IUPAC-namn 8-anilino-8-oxooctanoic acid
LEDER OC(=O)CCCCCCC(=O)Nc1ccccc1
Specifikationer
Kemiskt namn eller material Suberanilic Acid
Kvantitet 250 mg
InChI formel InChI=1S/C14H19NO3/c16-13(15-12-8-4-3-5-9-12)10-6-1-2-7-11-14(17)18/h3-5,8-9H,1-2,6-7,10-11H2,(H,15,16)(H,17,18)
Formel vikt 249.1365