Läs mer
Suberyl Glycine, TRC
Produktkod. 30132469
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132469 1 g
30004478 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132469 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6887551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60317-54-6
Molekylformel C10 H17 N O5
Molekylvikt (g/mol) 231.25
Synonym Octanoic acid, 8-[(carboxymethyl)amino]-8-oxo- (9CI, ACI), 8-[(Carboxymethyl)amino]-8-oxooctanoic acid (ACI), Suberylglycine, Suberyl glycine, N-Suberylglycine
IUPAC-namn 8-(carboxymethylamino)-8-oxooctanoic acid
LEDER OC(=O)CCCCCCC(=O)NCC(=O)O
Specifikationer
Kemiskt namn eller material Suberyl Glycine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C10H17NO5/c12-8(11-7-10(15)16)5-3-1-2-4-6-9(13)14/h1-7H2,(H,11,12)(H,13,14)(H,15,16)
Kvantitet 1 g
Formel vikt 231.1107
Procent renhet >95%