missing translation for 'onlineSavingsMsg'
Läs mer

MedChemExpress Substance P, Free Acid

Produktkod. 16499568
Encompass_Preferred
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16499568

Brand: MedChemExpress HYP1498

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Substance P, Free Acid is a native substance P analog, but shows no biological activity of substance P.

Specifikationer

CAS 71977-09-8
Innehåll och lagring Please store the product under the recommended conditions in the Certificate of Analysis.
För användning med (applikation) Neuroscience-Neuromodulation
Molekylvikt (g/mol) 1348.61 g/mol
Produkttyp Peptides
Kvantitet 10 mg
Frakt skick Room temperature
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.